2,3-Dimethoxybenzyl chloride 95%
Supplier: Apollo Scientific
Danger
OR200026-5G OR200026-1G OR200026-250MG OR200026-500MG
APOSOR200026-5GEA 236 EUR
APOSOR200026-5G APOSOR200026-1G APOSOR200026-250MG APOSOR200026-500MG
2,3-Dimethoxybenzyl chloride 95%
2,3-Dimethoxybenzyl chloride
Formula: C₉H₁₁ClO₂
Boiling Pt: 95 °C/1mmHg
Melting Pt: 69…71 °C
Density: 1.196 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD01310812
CAS Number: 3893-01-4