3-Methylxanthine
Supplier: Apollo Scientific
Warning
Synonyms: 3,7-Dihydro-3-methyl-1H-purine-2,6-dione, 2,6-Dihydroxy-3-methylpurine
OR20005-100G OR20005-250G OR20005-25G
APOSOR20005-100GEA 0 EUR
APOSOR20005-100G APOSOR20005-250G APOSOR20005-25G
3-Methylxanthine
3-Methylxanthine
Formula: C₆H₆N₄O₂
MW: 166.14 g/mol
Melting Pt: 360 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005580
CAS Number: 1076-22-8
EINECS: 214-058-1