3-Chloro-4-methylbenzoyl chloride 97%
Supplier: Apollo Scientific
Danger
OR200063-1G OR200063-250MG OR200063-5G OR200063-500MG
APOSOR200063-1GEA 510 EUR
APOSOR200063-1G APOSOR200063-250MG APOSOR200063-5G APOSOR200063-500MG
3-Chloro-4-methylbenzoyl chloride 97%
3-Chloro-4-methylbenzoyl chloride
Formula: C₈H₆Cl₂O
Boiling Pt: 135 °C/20mmHg
Storage Temperature: Ambient
MDL Number: MFCD07787492
CAS Number: 21900-30-1
UN: 1760
ADR: 8,III