Ethyl-3,4-diaminobenzoate
Supplier: Apollo Scientific
Warning
Synonyms: 3,4-Diaminobenzoic acid ethyl ester, Ethyl 3,4-diaminobenzoate
OR2001-100G OR2001-25G OR2001-5G
APOSOR2001-100GEA 119 EUR
APOSOR2001-100G APOSOR2001-25G APOSOR2001-5G
Ethyl-3,4-diaminobenzoate
Ethyl 3,4-diaminobenzoate
Formula: C₉H₁₂N₂O₂
MW: 180.21 g/mol
Melting Pt: 108…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00179337
CAS Number: 37466-90-3