(2R,6S)-2,6-Dimethyl-4-(3-nitropyridin-2-yl)morpholine
Supplier: Apollo Scientific
OR200145-1G OR200145-250MG OR200145-500MG OR200145-2.5G
APOSOR200145-1GEA 519 EUR
APOSOR200145-1G APOSOR200145-250MG APOSOR200145-500MG APOSOR200145-2.5G
(2R,6S)-2,6-Dimethyl-4-(3-nitropyridin-2-yl)morpholine
(2R,6S)-2,6-Dimethyl-4-(3-nitropyridin-2-yl)morpholine
Formula: C₁₁H₁₅N₃O₃
MW: 237.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28099707
CAS Number: 1706444-51-0