2-Methyl-8-quinolinol 98%
Supplier: Apollo Scientific
Warning
Synonyms: 8-Hydroxy-2-methylquinoline, 8-Hydroxyquinaldine, 2-Methyloxine
OR2004-100G OR2004-25G
APOSOR2004-100GEA 55.1 EUR
APOSOR2004-100G APOSOR2004-25G
2-Methyl-8-quinolinol 98%
2-Methyl-8-quinolinol
Formula: C₁₀H₉NO
MW: 159.19 g/mol
Boiling Pt: 267 °C
Melting Pt: 71…73 °C
Flash Pt: 139 °C
Storage Temperature: Ambient
MDL Number: MFCD00006765
CAS Number: 826-81-3
EINECS: 212-562-6
UN: 3077
ADR: 9+EHS III