Cyclohexyl methyl ketone 95%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Cyclohexylethanone
OR2012-25G OR2012-100G
APOSOR2012-25GEA 104 EUR
APOSOR2012-25G APOSOR2012-100G
Cyclohexyl methyl ketone 95%
Cyclohexyl methyl ketone
Formula: C₈H₁₄O
MW: 126.2 g/mol
Boiling Pt: 181…183 °C
Melting Pt: –34 °C
Density: 0.917 g/cm³ (25 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00040418
CAS Number: 823-76-7
EINECS: 212-517-0