2-Amino-5-bromobenzenesulphonamide
Supplier: Apollo Scientific
Warning
OR2016-1G OR2016-5G OR2016-10G
APOSOR2016-1GEA 21.9 EUR
APOSOR2016-1G APOSOR2016-5G APOSOR2016-10G
2-Amino-5-bromobenzenesulphonamide
2-Amino-5-bromobenzenesulphonamide
Formula: C₆H₇BrN₂O₂S
MW: 251.1 g/mol
Melting Pt: 170…176 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03426835
CAS Number: 54734-84-8