2,3,5,6-Tetrachloropyridine 98+%
Supplier: Apollo Scientific
Warning
OR2045-5G OR2045-1G
APOSOR2045-5GEA 29 EUR
APOSOR2045-5G APOSOR2045-1G
2,3,5,6-Tetrachloropyridine 98+%
2,3,5,6-Tetrachloropyridine
Formula: C₅HCl₄N
MW: 216.88 g/mol
Boiling Pt: 250…252 °C
Melting Pt: 88…93 °C
Flash Pt: 188 °C
Storage Temperature: Ambient
MDL Number: MFCD00092428
CAS Number: 2402-79-1
EINECS: 219-283-9
UN: 2811
ADR: 6.1 III