3,6-Dibromo-2-picoline 97%
Supplier: Apollo Scientific
Danger
Synonyms: 3,6-Dibromo-2-methylpyridine, 2,5-Dibromo-6-methylpyridine
OR2085-25G OR2085-5G OR2085-100G
APOSOR2085-25GEA 30.6 EUR
APOSOR2085-25G APOSOR2085-5G APOSOR2085-100G
3,6-Dibromo-2-picoline 97%
3,6-Dibromo-2-picoline
Formula: C₆H₅Br₂N
MW: 250.92 g/mol
Melting Pt: 34…38 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD06254589
CAS Number: 39919-65-8
UN: 2811
ADR: 6.1,III