3-Bromoanisole 99%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-3-methoxybenzene
OR2108-100G OR2108-250G OR2108-500G
APOSOR2108-100GEA 21.9 EUR
APOSOR2108-100G APOSOR2108-250G APOSOR2108-500G
3-Bromoanisole 99%
3-Bromoanisole
Formula: C₇H₇BrO
MW: 187.04 g/mol
Boiling Pt: 210…211 °C
Density: 1.477 g/cm³ (25 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00000081
CAS Number: 2398-37-0
EINECS: 219-264-5