2-(1-Piperidinyl)aniline
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-(1-Piperidino)aniline
OR21173-5G
OR21173-1G
OR21173-25G
APOSOR21173-5GEA
31.9
EUR
APOSOR21173-5G
APOSOR21173-1G
APOSOR21173-25G
2-(1-Piperidinyl)aniline
2-(1-Piperidinyl)aniline
Formula:
C₁₁H₁₆N₂ MW: 176.26 g/mol Boiling Pt: 96…98 °C/1mmHg Melting Pt: 44…47 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047467 CAS Number: 39643-31-7 UN: 2811 ADR: 6.1,III |