2-Bromo-4-nitroaniline
Supplier: Apollo Scientific
Warning
OR2118-5G OR2118-1G OR2118-25G
APOSOR2118-5GEA 36.4 EUR
APOSOR2118-5G APOSOR2118-1G APOSOR2118-25G
2-Bromo-4-nitroaniline
2-Bromo-4-nitroaniline
Formula: C₆H₅BrN₂O₂
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00025152
CAS Number: 13296-94-1