5,6-Dichloro-2-mercaptobenzimidazole 97%
Supplier: Apollo Scientific
Synonyms: 5,6-Dichloro-1H-benzimidazole-2-thiol
OR21183-5G
APOSOR21183-5GEA 119 EUR
APOSOR21183-5G
5,6-Dichloro-2-mercaptobenzimidazole 97%
5,6-Dichloro-2-mercaptobenzimidazole
Formula: C₇H₄Cl₂N₂S
MW: 219.09 g/mol
Melting Pt: 350 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00052398
CAS Number: 19462-98-7