4-Hydroxy-3,5-dimethylbenzaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 3,5-Dimethyl-4-hydroxybenzaldehyde
OR21230-100G OR21230-500G
APOSOR21230-100GEA 40.6 EUR
APOSOR21230-100G APOSOR21230-500G
4-Hydroxy-3,5-dimethylbenzaldehyde
4-Hydroxy-3,5-dimethylbenzaldehyde
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Melting Pt: 112…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00006946
CAS Number: 2233-18-3
EINECS: 218-774-5