5-Phenyl-2,4-pentadienoic acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Phenylpenta-2,4-dienoic acid
OR21264-25G
APOSOR21264-25GEA
154
EUR
APOSOR21264-25G
5-Phenyl-2,4-pentadienoic acid
5-Phenyl-2,4-pentadienoic acid
Formula:
C₆H₅CH=CHCH=CHCHCO₂H MW: 174.20 g/mol Melting Pt: 163…167 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014018 CAS Number: 1552-94-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...