2-Amino-4,6-dichlorophenol 95%
Supplier: Apollo Scientific
Warning
Synonyms: 3,5-Dichloro-6-hydroxyaniline
OR21268-100G OR21268-25G
APOSOR21268-100GEA 348 EUR
APOSOR21268-100G APOSOR21268-25G
2-Amino-4,6-dichlorophenol 95%
2-Amino-4,6-dichlorophenol
Formula: C₆H₅Cl₂NO
MW: 178.01 g/mol
Melting Pt: 89…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00035766
CAS Number: 527-62-8
EINECS: 208-421-3
UN: 2811
ADR: 6.1 III