4-Bromo-o-toluidine
Supplier: Apollo Scientific
Warning
Synonyms: 2-Amino-5-bromotoluene, 4-Bromo-2-methylaniline
OR21308-100G OR21308-25G OR21308-500G
APOSOR21308-100GEA 42.2 EUR
APOSOR21308-100G APOSOR21308-25G APOSOR21308-500G
4-Bromo-o-toluidine
4-Bromo-o-toluidine
Formula: C₇H₈BrN
MW: 186.05 g/mol
Boiling Pt: 240 °C
Melting Pt: 56…60 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007825
CAS Number: 583-75-5
UN: 3335
ADR: 9,III