(2,6-Dimethylpyridin-4-yl)methanol 95%
Supplier: Apollo Scientific
Warning
Synonyms: 2,6-Dimethyl-4-(hydroxymethyl)pyridine
OR2135-250MG
APOSOR2135-250MGEA 63.8 EUR
APOSOR2135-250MG
(2,6-Dimethylpyridin-4-yl)methanol 95%
(2,6-Dimethylpyridin-4-yl)methanol
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Melting Pt: 102…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00956275
CAS Number: 18088-01-2