4-(1-Piperidinyl)aniline 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Piperidinoaniline
OR21361-25G
APOSOR21361-25GEA 115 EUR
APOSOR21361-25G
4-(1-Piperidinyl)aniline 97%
4-(1-Piperidinyl)aniline
Formula: C₁₁H₁₆N₂
MW: 176.26 g/mol
Boiling Pt: 140…142 °C/1mmHg
Melting Pt: 26…29 °C
Density: 1.5937 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00051688
CAS Number: 2359-60-6