4,5-Dibromo-2-phenylpyridazin-3(2H)-one
Supplier: Apollo Scientific
Warning
OR21545-1G OR21545-250MG
APOSOR21545-1GEA 36.4 EUR
APOSOR21545-1G APOSOR21545-250MG
4,5-Dibromo-2-phenylpyridazin-3(2H)-one
4,5-Dibromo-2-phenylpyridazin-3(2H)-one
Formula: C₁₀H₆Br₂N₂O
MW: 329.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00052907
CAS Number: 14305-08-9