5-Chloro-o-anisidine 98%
Supplier: Apollo Scientific
Danger
Synonyms: 5-Chloro-2-methoxyaniline, 5-Chloro-2-anisidine, 2-Amino-4-chloroanisole
OR21760-100G OR21760-500G
APOSOR21760-100GEA 111 EUR
APOSOR21760-100G APOSOR21760-500G
5-Chloro-o-anisidine 98%
5-Chloro-o-anisidine
Formula: C₇H₈ClNO
MW: 157.6 g/mol
Boiling Pt: 135 °C/8mmHg
Melting Pt: 79…84 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007777
CAS Number: 95-03-4
EINECS: 202-385-2
UN: 2233
ADR: 6.1 III