(3-Methoxyphenyl)acetonitrile
Supplier: Apollo Scientific
Danger
Synonyms: 3-Methoxybenzyl cyanide, 2-(3-Methoxyphenyl)acetonitrile
OR21798-25G OR21798-5G
APOSOR21798-25GEA 29 EUR
APOSOR21798-25G APOSOR21798-5G
(3-Methoxyphenyl)acetonitrile
(3-Methoxyphenyl)acetonitrile
Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 278…282 °C
Density: 1.082 g/cm³ (25 °C)
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00001910
CAS Number: 19924-43-7
EINECS: 243-428-5
UN: 3276
ADR: 6.1,III