3,4-Dimethyl-1,5-dihydro-2H-pyrrol-2-one
Supplier: Apollo Scientific
Warning
OR21805-1G OR21805-250MG OR21805-500MG OR21805-5G
APOSOR21805-1GEA 340 EUR
APOSOR21805-1G APOSOR21805-250MG APOSOR21805-500MG APOSOR21805-5G
3,4-Dimethyl-1,5-dihydro-2H-pyrrol-2-one
3,4-Dimethyl-1,5-dihydro-2H-pyrrol-2-one
Formula: C₆H₉NO
MW: 111.14 g/mol
Melting Pt: 119…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00111570
CAS Number: 4030-22-2