2-Chloro-3-methyl-5-nitropyridine 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Chloro-5-nitro-3-picoline
OR21846-1G
OR21846-5G
APOSOR21846-1GEA
21.9
EUR
APOSOR21846-1G
APOSOR21846-5G
2-Chloro-3-methyl-5-nitropyridine 95%
2-Chloro-3-methyl-5-nitropyridine
Formula:
C₆H₅ClN₂O₂ Melting Pt: 42…44 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00173686 CAS Number: 22280-56-4 EINECS: 244-889-5 UN: 2811 ADR: 6.1,III |