4-Hydroxy-3-nitropyridine
Supplier: Apollo Scientific
Warning
Synonyms: 3-Nitro-4-pyridon, 3-Nitro-4-pyridone, 3-Nitropyridin-4-ol, 3-Nitro-4-pyridinol
OR21872-100G OR21872-25G OR21872-1KG OR21872-5KG
APOSOR21872-100GEA 74.1 EUR
APOSOR21872-100G APOSOR21872-25G APOSOR21872-1KG APOSOR21872-5KG
4-Hydroxy-3-nitropyridine
4-Hydroxy-3-nitropyridine
Formula: C₅H₄N₂O₃
MW: 140.10 g/mol
Melting Pt: 280…285 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00128874
CAS Number: 5435-54-1