2-Heptylcyclopentan-1-one oxime
Supplier: Apollo Scientific
OR21896-1G OR21896-5G OR21896-10G OR21896-25G OR21896-50G
APOSOR21896-1GEA 119 EUR
APOSOR21896-1G APOSOR21896-5G APOSOR21896-10G APOSOR21896-25G APOSOR21896-50G
2-Heptylcyclopentan-1-one oxime
2-Heptylcyclopentan-1-one oxime
Formula: C₁₂H₂₃NO
Storage Temperature: Ambient
MDL Number: MFCD09998437
CAS Number: 165385-93-3