4'-Butylacetophenone
Supplier: Apollo Scientific
Warning
Synonyms: 4'-n-Butylacetophenone, 4-Butylacetophenone, p-Butylacetophenone
OR21897-100G OR21897-25G OR21897-5G
APOSOR21897-100GEA 142 EUR
APOSOR21897-100G APOSOR21897-25G APOSOR21897-5G
4'-Butylacetophenone
4'-Butylacetophenone
Formula: C₁₂H₁₆O
Boiling Pt: 101…102 °C/1.5mmHg
Density: 0.957 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00017500
CAS Number: 37920-25-5
EINECS: 253-715-7