4-Propylcyclohexanol (cis- and trans- mixture)
Supplier: Apollo Scientific
Synonyms: 4-Propylcyclohexan-1-ol, 4-Propylcyclohexanol
OR21917-1G OR21917-250MG OR21917-5G OR21917-10G OR21917-25G
APOSOR21917-1GEA 74.1 EUR
APOSOR21917-1G APOSOR21917-250MG APOSOR21917-5G APOSOR21917-10G APOSOR21917-25G
4-Propylcyclohexanol (cis- and trans- mixture)
4-Propylcyclohexanol (cis- and trans- mixture)
Formula: C₉H₁₈O
MW: 142.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00091618
CAS Number: 52204-65-6