1-Bromo-4-hexylbenzene 97%
Supplier: Apollo Scientific
Warning
Synonyms: (4-Bromophenyl)hexane
OR21925-100G OR21925-25G
APOSOR21925-100GEA 313 EUR
APOSOR21925-100G APOSOR21925-25G
1-Bromo-4-hexylbenzene 97%
1-Bromo-4-hexylbenzene
Formula: C₁₂H₁₇Br
MW: 241.17 g/mol
Boiling Pt: 145 °C/0.75mmHg
Density: 1.18 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00061114
CAS Number: 23703-22-2