4'-tert-Butylacetophenone 95%
Supplier: Apollo Scientific
Warning
Synonyms: p-tert-Butylacetophenone
OR21926-25G OR21926-5G
APOSOR21926-25GEA 79.9 EUR
APOSOR21926-25G APOSOR21926-5G
4'-tert-Butylacetophenone 95%
4'-tert-Butylacetophenone
Formula: C₁₂H₁₆O
MW: 176.26 g/mol
Boiling Pt: 107…108 °C/5mmHg
Melting Pt: 17…18 °C
Density: 0.964 g/cm³ (25 °C)
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00017256
CAS Number: 943-27-1
EINECS: 213-399-3
UN: 1224
ADR: 3,III