1,8-Bis(dimethylamino)naphthalene 98%
Supplier: Apollo Scientific
Warning
Synonyms: N1,N1,N8,N8-Tetramethylnaphthalene-1,8-diamine
OR21927-10G OR21927-25G OR21927-100G
APOSOR21927-10GEA 21.9 EUR
APOSOR21927-10G APOSOR21927-25G APOSOR21927-100G
1,8-Bis(dimethylamino)naphthalene 98%
1,8-Bis(dimethylamino)naphthalene
Formula: C₁₄H₁₈N₂
MW: 214.31 g/mol
Boiling Pt: 144…145 °C/7mmHg
Melting Pt: 49…51 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00003920
CAS Number: 20734-58-1
EINECS: 244-001-6