3,5-Di-tert-butyl-4-hydroxybenzonitrile 97%
Supplier: Apollo Scientific
|
Warning
|
OR21941-1G
OR21941-5G
APOSOR21941-1GEA
31.9
EUR
APOSOR21941-1G
APOSOR21941-5G
3,5-Di-tert-butyl-4-hydroxybenzonitrile 97%
3,5-Di-tert-butyl-4-hydroxybenzonitrile
Formula:
C₁₅H₂₁NO Melting Pt: 141…143 °C Storage Temperature: Ambient |
MDL Number:
MFCD00156137 CAS Number: 1988-88-1 UN: 3439 ADR: 6.1,III |