2,6-Di-tert-butyl-p-benzoquinone
Supplier: Apollo Scientific
Warning
Synonyms: 2,6-Di-tert-butylcyclohexa-2,5-diene-1,4-dione
OR21944-25G OR21944-5G
APOSOR21944-25GEA 87 EUR
APOSOR21944-25G APOSOR21944-5G
2,6-Di-tert-butyl-p-benzoquinone
2,6-Di-tert-butyl-p-benzoquinone
Formula: C₁₄H₂₀O₂
Boiling Pt: 75…80 °C/0.7mmHg
Melting Pt: 65…67 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001601
CAS Number: 719-22-2