1-Pentyl-4-ethynylbenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-N-Pentylphenylacetylene, 4-n-Pentylphenylacetylene, (4-Pentylphenyl)acetylene, 1-Ethynyl-4-pentylbenzene, (4-Pentylphenyl)ethyne
OR21955-100G
APOSOR21955-100GEA 166 EUR
APOSOR21955-100G
1-Pentyl-4-ethynylbenzene 98%
1-Pentyl-4-ethynylbenzene
Formula: C₁₃H₁₆
MW: 172.27 g/mol
Boiling Pt: 172 °C
Density: 0.885 g/cm³ (25 °C)
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00173884
CAS Number: 79887-10-8