1-Butyl-4-ethynylbenzene
Supplier: Apollo Scientific
Warning
Synonyms: 4-Butylphenylacetylene
OR21956-25G OR21956-5G
APOSOR21956-25GEA 87 EUR
APOSOR21956-25G APOSOR21956-5G
1-Butyl-4-ethynylbenzene
1-Butyl-4-ethynylbenzene
Formula: C₁₂H₁₄
MW: 158.24 g/mol
Boiling Pt: 70…71 °C
Density: 0.906 g/cm³ (25 °C)
Flash Pt: 101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00173885
CAS Number: 79887-09-5