1-Propyl-4-ethynylbenzene
Supplier: Apollo Scientific
Warning
Synonyms: 1-Ethynyl-4-propylbenzene, (4-Propylphenyl)acetylene, 4-n-Propylphenylacetylene, (4-Propylphenyl)ethyne
OR21957-100G
APOSOR21957-100GEA 127 EUR
APOSOR21957-100G
1-Propyl-4-ethynylbenzene
1-Propyl-4-ethynylbenzene
Formula: C₁₁H₁₂
MW: 144.22 g/mol
Boiling Pt: 205 °C
Density: 0.91 g/cm³ (25 °C)
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00173886
CAS Number: 62452-73-7