2-(5-Methyl-2-phenyl-1,3-oxazol-4-yl)acetic acid 95%
Supplier: Apollo Scientific
Warning
OR21964-1G OR21964-250MG
APOSOR21964-1GEA 43.5 EUR
APOSOR21964-1G APOSOR21964-250MG
2-(5-Methyl-2-phenyl-1,3-oxazol-4-yl)acetic acid 95%
2-(5-Methyl-2-phenyl-1,3-oxazol-4-yl)acetic acid
Formula: C₁₂H₁₁NO₃
Melting Pt: 131…133 °C
Storage Temperature: Ambient
MDL Number: MFCD00100005
CAS Number: 107367-98-6