Methyl-2-bromo-5-chlorobenzoate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 2-bromo-5-chlorobenzoate
OR2198-100G OR2198-25G
APOSOR2198-100GEA 114 EUR
APOSOR2198-100G APOSOR2198-25G
Methyl-2-bromo-5-chlorobenzoate 98%
Methyl 2-bromo-5-chlorobenzoate
Formula: C₈H₆BrClO₂
MW: 249.49 g/mol
Melting Pt: 37…40 °C
Storage Temperature: Ambient
MDL Number: MFCD00144763
CAS Number: 27007-53-0