Methyl-3-chloro-5-iodobenzoate
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 3-chloro-5-iodobenzoate
OR2199-5G
APOSOR2199-5GEA 60.9 EUR
APOSOR2199-5G
Methyl-3-chloro-5-iodobenzoate
Methyl 3-chloro-5-iodobenzoate
Formula: C₈H₆ClIO₂
MW: 296.49 g/mol
Melting Pt: 41…43 °C
Storage Temperature: Ambient
MDL Number: MFCD00673018
CAS Number: 289039-85-6