Isoquinoline-3-amine 97%
Supplier: Apollo Scientific
Warning
Synonyms: Isoquinolin-3-amine, 3-Aminoisoquinoline
OR22000-1G OR22000-10G OR22000-5G
APOSOR22000-1GEA 26.1 EUR
APOSOR22000-1G APOSOR22000-10G APOSOR22000-5G
Isoquinoline-3-amine 97%
Isoquinoline-3-amine
Formula: C₉H₈N₂
MW: 144.1700 g/mol
Melting Pt: 174…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00102190
CAS Number: 25475-67-6