p-tert-Butylanisole 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4-tert-Butylanisole, 1-(tert-Butyl)-4-methoxybenzene
OR2202-100G
APOSOR2202-100GEA 87 EUR
APOSOR2202-100G
p-tert-Butylanisole 97%
p-tert-Butylanisole
Formula: C₁₁H₁₆O
MW: 164.25 g/mol
Boiling Pt: 222 °C
Melting Pt: 18 °C
Density: 0.94 g/cm³ (25 °C)
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00026309
CAS Number: 5396-38-3
EINECS: 226-412-2