4-Butylbiphenyl 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Butyl-1,1'-Biphenyl
OR22067-100G OR22067-25G OR22067-500G
APOSOR22067-100GEA 56.7 EUR
APOSOR22067-100G APOSOR22067-25G APOSOR22067-500G
4-Butylbiphenyl 98%
4-Butylbiphenyl
Formula: C₁₆H₁₈
MW: 210.32 g/mol
Boiling Pt: 318…319 °C
Melting Pt: 44414 °C
Density: 0.953 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00100665
CAS Number: 37909-95-8