4,6-Di(tert-butyl)benzene-1,3-diol 95%
Supplier: Apollo Scientific
Warning
Synonyms: 4,6-Di-tert-butylresorcinol, 4,6-Di-tert-butylbenzene-1,3-diol
OR22090-1G OR22090-5G
APOSOR22090-1GEA 132 EUR
APOSOR22090-1G APOSOR22090-5G
4,6-Di(tert-butyl)benzene-1,3-diol 95%
4,6-Di(tert-butyl)benzene-1,3-diol
Formula: C₁₄H₂₂O₂
MW: 222.33 g/mol
Melting Pt: 120…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00044513
CAS Number: 5374-06-1
EINECS: 226-366-3