Pyrrolidine-2-thione
Supplier: Apollo Scientific
Warning
OR22113-1G OR22113-5G
APOSOR22113-1GEA 94.4 EUR
APOSOR22113-1G APOSOR22113-5G
Pyrrolidine-2-thione
Pyrrolidine-2-thione
Formula: C₄H₇NS
Melting Pt: 110…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00049351
CAS Number: 2295-35-4