7-Hydroxy-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine
Supplier: Apollo Scientific
Warning
Synonyms: 7-Hydroxy-5-methyl-1,3,4-triazaindolizine, 5-Methyl-[1,2,4]triazolo[1,5-a]pyrimidin-7-ol, 4-Hydroxy-6-methyl-1,3,3a,7-tetraazaindene
OR22121-25G OR22121-5G
APOSOR22121-25GEA 29 EUR
APOSOR22121-25G APOSOR22121-5G
7-Hydroxy-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine
7-Hydroxy-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine
Formula: C₆H₆N₄O
MW: 150.14 g/mol
Melting Pt: 280…283 °C
Storage Temperature: Ambient
MDL Number: MFCD00005555
CAS Number: 2503-56-2
EINECS: 219-706-7