3(2H)-Pyridazinone 95%
Supplier: Apollo Scientific
Warning
Synonyms: 2,3-dihydropyridazin-3-one, Pyridazin-3(2H)-one
OR2215-25G OR2215-5G OR2215-100G
APOSOR2215-25GEA 65.4 EUR
APOSOR2215-25G APOSOR2215-5G APOSOR2215-100G
3(2H)-Pyridazinone 95%
3(2H)-Pyridazinone
Formula: C₄H₄N₂O
MW: 96.0874 g/mol
Boiling Pt: 101 °C/1.5mmHg
Melting Pt: 98…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00099605
CAS Number: 504-30-3