2,5-Dibromoterephthalic acid
Supplier: Apollo Scientific
Danger
Synonyms: 2,5-Dibromoterephtalic acid
OR22280-1G OR22280-5G OR22280-10G
APOSOR22280-1GEA 21.9 EUR
APOSOR22280-1G APOSOR22280-5G APOSOR22280-10G
2,5-Dibromoterephthalic acid
2,5-Dibromoterephthalic acid
Formula: C₈H₄Br₂O₄
MW: 323.9200 g/mol
Melting Pt: 318 °C
Storage Temperature: Ambient
MDL Number: MFCD00204593
CAS Number: 13731-82-3
UN: 1759
ADR: 8,III