1-(2-Chloro-6-hydroxyphenyl)ethanone 97+%
Supplier: Apollo Scientific
OR22395-1G OR22395-5G OR22395-25G
APOSOR22395-1GEA 63.8 EUR
APOSOR22395-1G APOSOR22395-5G APOSOR22395-25G
1-(2-Chloro-6-hydroxyphenyl)ethanone 97+%
1-(2-Chloro-6-hydroxyphenyl)ethanone
Formula: C₈H₇ClO₂
MW: 170.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00218550
CAS Number: 55736-04-4