1-(4-Nitrophenyl)glycerol 99%
Supplier: Apollo Scientific
Warning
Synonyms: PNPG, 1-(4-Nitrophenyl)-1,2,3-propanetriol
OR22490-1G OR22490-250MG OR22490-5G
APOSOR22490-1GEA 0 EUR
APOSOR22490-1G APOSOR22490-250MG APOSOR22490-5G
1-(4-Nitrophenyl)glycerol 99%
1-(4-Nitrophenyl)glycerol
Formula: C₉H₁₁NO₅
MW: 213.19 g/mol
Melting Pt: 93…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00017037
CAS Number: 2207-68-3
EINECS: 218-624-9